Would you like to log in again?
KpnI
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R6341 R6345
PAR6341EA 114.87 USD InStock
PAR6341 PAR6345
KpnI
Nucleic Acid Reagents Restriction Enzymes
G GTAC/C C\CATG G
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty