Would you like to log in again?
Cla I (Blue/White Cloning and Genome Qualified), Promega
Supplier: Promega
R6555 R6551
PAR6551EA 118.75 USD InStock
PAR6551 PAR6555
Cla I (Blue/White Cloning and Genome Qualified), Promega
Nucleic Acid Reagents Restriction Enzymes
AT/CG AT TA GC\TA
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty