Would you like to log in again?
AttoPhos® Substrate, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S1011 S1013 S1022 S1012
PAS1012EA 243.3 USD InStock
PAS1012 PAS1013 PAS1011 PAS1022
AttoPhos® Substrate, Promega
Assays Protein Assays
A highly sensitive, flluorescent alkaline phosphatase substrate.
The AttoPhos® Substrate (2´-[2-benzothiazoyl]-6´-hydroxybenzothiazole phosphate [BBTP]) is cleaved by alkaline phosphatase (AP) to produce inorganic phosphate (Pi) and the alcohol, 2´-[2- benzothiazoyl]-6´- hydroxybenzothiazole (BBT). This enzyme-catalyzed conversion of AttoPhos® Substrate to BBT is accompanied by an enhancement in fluorescence properties that include increased quantum efficiency plus fluorescence excitation and emission maxima that are red-shifted further into the visible region.
The AttoPhos® Buffer is optimized for sensitive detection of alkaline phosphatase (AP) when used to prepare working solutions with AttoPhos® Substrate
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty