Would you like to log in again?
PDE-Glo Phosphodiesterase Assay, Promega
Supplier: Promega
PDE-Glo™
V1361
PAV1361EA 915.48 USD InStock
PAV1361
PDE-Glo Phosphodiesterase Assay, Promega
Assays Enzyme Assays
The PDE-Glo Phosphodiesterase Assay is a luminescent, high-throughput screening method for measuring cyclic nucleotide phosphodiesterase activity from purified sources.
  • Detects Cyclic Nucleotide Phosphodiesterase Activity
  • Assay complete in >1 hour after PDE reaction
  • Scale from 1-tube to 1536-well plate formats
  • Simple luminescence detection
The PDE-Glo Phosphodiesterase Assay is a luminescent, high-throughput screening (HTS) method for measuring cyclic nucleotide phosphodiesterase activity from purified sources. Cyclic nucleotide phosphodiesterases (PDEs) are involved in a myriad of cellular processes due to their ability to hydrolyze, and thus control, the levels of the second-messenger signaling molecules cAMP and cGMP. The availability of selective inhibitors for PDEs has facilitated their use as tools to study cyclic nucleotide signaling and paved the way to investigate the role of PDEs in cellular and tissue pathologies. The PDE-Glo Phosphodiesterase Assay allows lead candidates to be identified from compound libraries. The assay is designed for 384-well plates, but assay volumes can easily be scaled for 96- or 1536-well plates. The PDE-Glo Phosphodiesterase Assay is optimized to work with both cAMP- and cGMP-dependent phosphodiesterases. The total time required for the assay from start to finish is less than 1 hour after the PDE reaction is complete.
The assay is optimized to work with both cAMP- and cGMP-dependent phosphodiesterases. Total time required for the assay is less than one hour after the PDE reaction is complete.
The PDE-Glo™ assay allows lead candidate identification from compound libraries. Designed for 384-well plates, the assay volumes can be easily scaled for 96 or 1536-well plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty