Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml, Promega

Supplier: Promega
V1701
PAV1701EA 362.26 USD InStock
PAV1701
PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml, Promega
Assays Protein Assays
The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are also part of the PI3K-Glo kinase profiling system.

  • Phosphoinositide-3 KInase (PI3K) Substrates
  • Use in screening assays with purified PI3K Class I lipid kinases and ADP-Glo™ Kinase Assay
  • PIP2:3PS supplied at 0.25mg in 0.25ml
  • PI:3PS supplied at 0.5mg in 0.5ml

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are part of the PI3K-Glo kinase profiling system. Promega lipid kinase enzymes, substrates and detection systems provide a complete set of reagents for performing phosphoinositide lipid kinase (PIK) reactions using a luminescent ADP-detection platform, the ADP-Glo Kinase Assay. The lipid kinase reaction is performed by incubating lipid substrate (PI:3PS or PIP2:3PS) with a recombinant enzyme and ATP, and the kinase activity is measured using the ADP-Glo Kinase Assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty