Would you like to log in again?
ADP-Glo Kinase Assay with PIP2:3PS, 1,000 assays, Promega
Supplier: Promega
V1791 V1792
PAV1791EA 1060.56 USD InStock
PAV1791 PAV1792
ADP-Glo Kinase Assay with PIP2:3PS, 1,000 assays, Promega
Assays Cellular Assays
The ADP-Glo Kinase assay and lipid kinase substrates PIP2:3PS and PI:3PS are components of the PI3K-Glo Class I Kinase Profiling System that are available for purchase separately.
  • Substrate and Assay Reagents for Detecting Class I PI3K Activity
  • Use in screening assays with purified PI3K Class I lipid kinases
  • Simple, scalable "add-and-read" assay format
The ADP-Glo Kinase assay and lipid kinase substrates PIP2:3PS and PI:3PS are components of the PI3K-Glo Class I Kinase Profiling System that are available for purchase separately. Promega lipid kinase enzymes, substrates and detection systems provide a complete set of reagents for performing phosphoinositide lipid kinase (PIK) reactions using a luminescent ADP-detection platform, the ADP-Glo Kinase Assay. The lipid substrates are supplied as frozen small unilamellar vesicles containing a mixture of phosphatidylinositol (PI) or phosphoinositol-4,5-bisphosphate (PIP2) at a 1:3 ratio with phosphatidylserine (PS) as carrier lipid. A substrate composed of PIP2 and PS at a 1:3 ratio was optimized to use with class I PI3Ks. The lipid kinase reaction is performed by incubating lipid substrate (PI:3PS or PIP2:3PS) with a recombinant enzyme and ATP, and the kinase activity is measured using the ADP-Glo Kinase Assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty