Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose, Low Melting Point, Analytical Grade, Promega Corporation, Promega

Supplier: Promega
V2111
PAV2111EA 195.52 USD InStock
PAV2111
Agarose, Low Melting Point, Analytical Grade, Promega Corporation, Promega
Electrophoresis Gels Agarose and Acrylamide
Agarose for applications that require recovery of intact DNA fragments after gel electrophoresis.

  • Quality tested and certified free of DNase and RNase activity
  • Form: white powder
  • Gelling point (1,5%): 24 to 28 °C
  • Melting point (1,5%): ≤65,5 °C
  • Sulphate:≤0,12%
  • EEO (–mr): ≤0,11
  • Gel strength (1%): ≥300g/cm²

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty