Would you like to log in again?
Serine/Threonine Phosphatase Assay System, 96 reactions, Promega
Supplier: Promega
V2460
PAV2460EA 719.24 USD InStock
PAV2460
Serine/Threonine Phosphatase Assay System, 96 reactions, Promega
Assays Enzyme Assays
The Non-Radioactive Phosphatase Assay Systems provide a fast, convenient and flexible alternative for measuring protein phosphatase activity.
  • Absorbance-Based Detection of Phosphatase Activity
  • Choose from serine/threonine or tyrosine phosphatase assays
  • Use with purified samples, crude cell or tissue extracts
  • Convenient alternative to radioactive phosphatase assays
The Non-Radioactive Phosphatase Assay Systems provide a fast, convenient and flexible alternative for measuring protein phosphatase activity. These systems determine the amount of free phosphate generated in a reaction by measuring the absorbance of a molybdate:malachite green:phosphate complex. These systems allow the use of a variety of buffer conditions and substrates, including naturally phosphorylated proteins or synthetic phosphopeptides. The Serine/Threonine Phosphatase Assay System contains the chemically synthesized phosphopetide, RRA(pT)VA, a peptide substrate that is compatible with several serine/threonine phosphatases such as the Protein Phosphatases 2A, 2B, and 2C. However the supplied phosphopeptide is a poor substrate for Protein Phosphatase 1 because of its more stringent structural requirements. The Tyrosine Phosphatase Assay System contains two chemically synthesized phosphopeptides, END(pY)INASL and DADE(pY)LIPQQG, that serve as substrates for many protein tyrosine phosphatases. The effective range for the detection of phosphate released during an assay using the Phosphatase Assay Systems is 100-4,000pmol of phosphate. In addition to measuring phosphatase activity in partially fractionated and purified samples, the Phosphatase Assay Systems can also measure phosphatase activity in crude cell or tissue extracts. For this application, the high concentration of phosphate in these preparations is eliminated prior to performing the assay using the supplied Spin Columns, which rapidly and effectively remove free phosphate and other low-molecular-weight inhibitors from the sample. In addition, a unique Molybdate Dye Additive that is combined with the Molybdate Dye Solution aids in the solubilization of proteins exposed to the acid conditions of the Molybdate Dye Solution, which alone could potentially cause precipitation of the proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty