Would you like to log in again?
IRAK4 Kinase Enzyme System, Promega
Supplier: Promega
V2621 V2622
PAV2621EA 660.77 USD InStock
PAV2621 PAV2622
IRAK4 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human IRAK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Interleukin-1 receptor-associated kinase 4 is an important mediator in the signal transduction of Toll-like receptor and IL1R family members.
  • Easily Screen and Profile IRAK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human IRAK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Interleukin-1 receptor-associated kinase 4 (IRAK4) is an important mediator in the signal transduction of Toll-like receptor (TLR) and IL1R family members. IRAK4 is involved in the Toll-like receptor signaling pathway leading to Apoptosis. IRAK4 has molecular functions like protein binding, ATP binding, kinase activity and magnesium ion binding. Toll/IL-1 receptor family members like IRAK4 are central components of host defense mechanisms in a variety of species. One well conserved element in their signal transduction is the Ser/Thr kinase activity, which couple early signaling events in a receptor complex at the plasma membrane to larger signalosomes in the cytosol. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: IRAK4, 10ug (Human, recombinant full-length). MW: ~81kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. IRAK4 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/51135/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty