Would you like to log in again?
Guanidine Thiocyanate
Supplier: Promega
V2791
PAV2791EA 64.44 USD InStock
PAV2791
Guanidine Thiocyanate
Enzymes
Protein denaturant used most commonly for the isolation of intact RNA due to its ability to inhibit RNase.
  • Quality tested and certified free of DNase, RNase and protease activity
Guanidine Thiocyanate, Molecular Grade, at high concentrations, is a protein denaturant used most commonly for the isolation of intact RNA due to its ability to inhibit RNase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty