Would you like to log in again?
PLK1 Kinase Enzyme System
Supplier: Promega
V2842 V2841
PAV2841EA 624.55 USD InStock
PAV2841 PAV2842
PLK1 Kinase Enzyme System
Assays Enzyme Assays
Recombinant full-length human PLK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. A member of the Polo-Like Kinase family, PLK1 localizes to centrosomes and undergoes dramatic subcellular relocation during the cell cycle.
  • Easily Screen and Profile PLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PLK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. PLK1 is a member of the Polo-Like Kinase family, which localizes to centrosomes or spindle pole bodies and undergoes dramatic subcellular relocation during the cell cycle. Deregulated activities of PLKs often result in abnormalities in centrosome duplication, maturation and/or microtubule dynamics. PLKs also regulate the function of the Golgi complex. Deregulated expression of human PLK1 is strongly correlated with the development of many types of malignancies, and ectopic expression of PLK1 dominant negative protein leads to rapid cell death. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PLK1, 10ug (Human, recombinant full-length). MW: ~70kDa. Substrate: Casein, Dephosphorylated (Bovine). Other: Reaction Buffer, DTT. PLK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5347/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty