Would you like to log in again?
CSK Kinase Enzyme System, Promega
Supplier: Promega
V2981 V2982
PAV2981EA 588.87 USD InStock
PAV2981 PAV2982
CSK Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human CSK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CSK has been shown to downregulate the tyrosine kinase activity of c-src through tyrosine phosphorylation of the c-src carboxy terminus.
  • Easily Screen and Profile CSK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human CSK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CSK is a cytoplasmic tyrosine kinase that has been shown to downregulate the tyrosine kinase activity of c-src through tyrosine phosphorylation of the c-src carboxy terminus. A yeast 2-hybrid system has been used to identify proteins associated with CSK. The Src homology-3 (SH3) domain of CSK associates with a proline-rich region of PEP, a protein-tyrosine phosphatase expressed in hemopoietic cells. This association is highly specific, and it is speculated that PEP may be an effector and/or regulator of CSK in T cells and other hemopoietic cells. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CSK, 10ug (Human, recombinant full-length). MW: ~78kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. CSK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1445/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty