Would you like to log in again?
GSK3 alpha Kinase Enzyme System, Promega
Supplier: Promega
V3052 V3051
PAV3052EA 8648.06 USD InStock
PAV3052 PAV3051
GSK3 alpha Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human GSK3 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 alpha is a multifunctional protein serine kinase implicated in the control of several regulatory proteins.
  • Easily Screen and Profile GSK3α Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human GSK3 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 alpha is a multifunctional protein serine kinase, homologous to Drosophila 'shaggy' (zeste-white3) and implicated in the control of several regulatory proteins including glycogen synthase and transcription factors (e.g., JUN). GSK3 alpha also plays a role in the WNT and PI3K signaling pathways. Alzheimer disease is associated with increased production and aggregation of amyloid-beta-40 and -42 peptides into plaques. GSK3 alpha is required for maximal production of the beta-amyloid-40 and -42 peptides generated from the amyloid precursor protein by presenilin-dependent gamma-secretase cleavage. In vitro, lithium, a GSK3 alpha inhibitor, blocks the production of the beta-amyloid peptides by interfering with the gamma-secretase step. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: GSK3 alpha, 10ug (Human, recombinant full-length). MW: ~81kDa. Substrate: GSK3 Substrate (YRRAAVPPSPSLSRHSSPHQ(pS)EDEEE); derived from human muscle glycogen synthase 1 (amino acid 636-661). Other: Reaction Buffer, DTT. GSK3 alpha NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2931/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty