Would you like to log in again?
HER4 Kinase Enzyme System, Promega
Supplier: Promega
V3102 V3101
PAV3102EA 8821.02 USD InStock
PAV3102 PAV3101
HER4 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human HER4 (amino acids 682-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER4 or ERBB4 is a transmembrane receptor tyrosine kinase that belongs to the epidermal growth factor receptor family.
  • Easily Screen and Profile HER4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human HER4 (amino acids 682-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER4 or ERBB4 is a transmembrane receptor tyrosine kinase that belongs to the epidermal growth factor receptor family. Through its interaction with its activating ligand, heregulin, HER4 can regulate proliferation and differentiation of cells. HER4 ectodomain is cleaved by a metalloprotease, gamma-secretase, that releases the HER4 intracellular domain from the membrane and facilitates its translocation to the nucleus. The kinase activity of HER4 is both necessary and sufficient to trigger an antiproliferative response in human breast cancer cells. Increased expression of HER4 has been detected in various cancers, which usually correlates to better survival. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: HER4, 10ug (Human, recombinant; amino acids 682-993). MW: ~57kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. HER4 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2066/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty