Would you like to log in again?
Acrylamide, Molecular Grade, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V3115
PAV3115EA 192.4 USD InStock
PAV3115
Acrylamide, Molecular Grade, Promega
Electrophoresis Gels Agarose and Acrylamide
Used for the electrophoretic separation of nucleic acids and proteins.
  • Quality tested and certified free of DNase, RNase and protease activity
Acrylamide, Molecular Grade, is used for the electrophoretic separation of nucleic acids and proteins. Very small DNA fragments, such as those generated by sequencing reactions, can be resolved by polyacrylamide gel electrophoresis. Proteins can be separated by a variety of techniques, including denaturing gel electrophoresis using SDS or urea, isoelectric focusing and native gel electrophoresis in a wide variety of buffers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty