Would you like to log in again?
PAK4 Kinase Enzyme System, Promega
Supplier: Promega
V3201 V3202
PAV3201EA 612.3 USD InStock
PAV3201 PAV3202
PAK4 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human PAK4 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. PAK4 is a member of the p21-activated kinases (PAKs), which have been implicated in the regulation of cell morphology, motility and transformation.
  • Easily Screen and Profile PAK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PAK4 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. PAK4 is a recently identified member of the p21-activated kinases (PAKs), which have been implicated in the regulation of cell morphology, motility and transformation. These serine/threonine kinases are activated by and are effectors of small GTPases, cdc 42 and Rac. PAK4 belongs to the Group II PAKs that also includes PAK5 and PAK6. PAK4 has been shown to regulate cell morphology and cytoskeletal organization in mammalian cells. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PAK4, 10ug (Human, recombinant full-length). MW: ~90kDa. Substrate: Modified AKT Substrate II peptide (modified-CKRPRAASFAE); based on the N-terminus of GSK3. Other: Reaction Buffer, DTT. PAK4 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/10298/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty