Would you like to log in again?
EPHA1 Kinase Enzyme System, Promega
Supplier: Promega
V3562 V3561
PAV3562EA 8648.06 USD InStock
PAV3562 PAV3561
EPHA1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant mouse EPHA1 (amino acids 569-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events.
  • Easily Screen and Profile EPHA1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant mouse EPHA1 (amino acids 569-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the Eph subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. EPHA1 seems to be a marker of the differentiated normal epidermis, and its downregulation in nonmelanoma skin cancer may contribute to carcinogenesis of these very frequent human tumors. EPHA1 represents a new potential prognostic marker and therapeutic target in nonmelanoma skin cancer. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: EPHA1, 10ug (Human, recombinant; amino acids 569-end). MW: ~71kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. EPHA1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2041/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty