Would you like to log in again?
Agarose
Supplier: Promega
V3841
PAV3841EA 184.05 USD InStock
PAV3841
Agarose
Enzymes
Premium agarose used for isolating DNA fragments from 10 to 1,000bp.
  • Quality tested and certified free of DNase and RNase activity
Agarose, LMP, Preparative Grade for Small Fragments (10 to 1,000bp), is a premium agarose used for isolating DNA fragments from 10 to 1,000bp. The isolated DNA fragments can be used in various molecular biology applications: 1) restriction digestion, 2) ligation and transformation, and 3) random prime labeling. LMP = low melting point (i.e., </=65 degrees C).
LMP stands for Low Melting Point.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty