Would you like to log in again?
HER2 Kinase Enzyme System, Promega
Supplier: Promega
V3891 V3892
PAV3892EA 8648.06 USD InStock
PAV3892 PAV3891
HER2 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human HER2 (amino acids 676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER2 gene encodes a cell-surface glycoprotein tyrosine kinase receptor with extensive homology to epidermal growth factor receptor.
  • Easily Screen and Profile HER2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human HER2 (amino acids 676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER2 gene encodes a cell-surface glycoprotein tyrosine kinase receptor with extensive homology to the epidermal growth factor receptor. HER2 is an oncogene, and overexpression of unaltered HER2 coding sequences in NIH/3T3 cells results in cellular transformation and tumorigenesis. HER2 is amplified in about 30% of primary human breast malignancies, and overexpression of HER2 is associated with the most aggressive tumors that show uncontrolled proliferation, resistance to apoptosis and increased motility. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: HER2, 10ug (Human, recombinant; amino acids 676-end). MW: ~116kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT. HER2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2064/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty