Would you like to log in again?
AXL Kinase Enzyme System, Promega
Supplier: Promega
V3962 V3961
PAV3962EA 8648.06 USD InStock
PAV3962 PAV3961
AXL Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human AXL (amino acids 473-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. AXL is a member of the receptor tyrosine kinase family, which has oncogenic potential and is implicated in human myeloid leukemia.
  • Easily Screen and Profile AXL Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human AXL (amino acids 473-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. AXL is a member of the receptor tyrosine kinase family, which has oncogenic potential and is implicated in human myeloid leukemia. AXL is a member of a complex signaling network that is involved in the control of cell proliferation and differentiation. Overexpression of AXL cDNA in NIH/3T3 cells induces neoplastic transformation of these cells with the concomitant appearance of a 140kDa AXL tyrosine-phosphorylated protein. Expression of AXL cDNA in the baculovirus system results in the expression of the appropriate recombinant protein that is recognized by anti-phosphotyrosine antibodies, confirming that the AXL protein is tyrosine phosphorylated. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AXL, 10ug (Human, recombinant; amino acids 473-end). MW: ~55kDa. Substrate: Axltide (KKSRGDYMTMQIG); derived from the mouse Insulin receptor substrate 1 (amino acid 979-989). Other: Reaction Buffer, DTT. AXL NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/558/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty