Would you like to log in again?
AKT3 Kinase Enzyme System, Promega
Supplier: Promega
V4010 V6271
PAV4010EA 572.21 USD InStock
PAV4010 PAV6271
AKT3 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human AKT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT3 or Protein Kinase B gamma (PKBgamma) is a serine/threonine kinase that is a member of the AKT family.
  • Easily Screen and Profile AKT3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human AKT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT3 or Protein Kinase B gamma (PKBgamma) is a serine/threonine kinase that is a member of the AKT family. AKT3 is activated in cells exposed to diverse stimuli such as hormones, growth factors and extracellular matrix components (1). AKT3 phosphorylates and regulates the function of many cellular proteins involved in processes that include cellular metabolism, survival/apoptosis and proliferation. Recent evidence indicates that AKT3 is frequently overexpressed in many types of human cancers including breast and prostate (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AKT3, 10ug (Human, recombinant full-length). MW: ~85kDa. Substrate: Akt (SGK) substrate peptide (RPRAATF); derived from the N-terminus of GSK3. Other: Reaction Buffer, DTT. AKT3 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/10000/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty