Would you like to log in again?
AMPK (A2/B1/G1) Kinase Enzyme System, Promega
Supplier: Promega
V6275 V4014
PAV4014EA 1081.71 USD InStock
PAV4014 PAV6275
AMPK (A2/B1/G1) Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human AMPK (A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for diabetes treatment.
  • Easily Screen and Profile AMPK (A2/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human AMPK (combination of A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for the treatment of diabetes (1). AMPK is viewed as a fuel sensor for glucose and lipid metabolism by modulating the activity of the autonomous nervous system in vivo. Short-term overexpression of a constitutively active form of AMPK in the liver leads to mild hypoglycemia and fatty liver due to increased fatty acid utilization (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AMPK (A2/B1/G1), 10ug (Human, recombinant full-length). MW: ~69kDa (A2); ~38kDa (B1); ~40kDa (G1). Substrate: SAMStide (HMRSAMSGLHLVKRR); derived from the mouse acetyl-Coenzyme A carboxylase alpha (amino acids 73-85). Other: Reaction Buffer, DTT, AMP Solution. AMPK (A2/B1/G1) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5563/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty