Would you like to log in again?
ADP-Glo Kinase Assay+ MAPKAPK3 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4027
PAV4027EA 1140.69 USD InStock
PAV4027
ADP-Glo Kinase Assay+ MAPKAPK3 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human MAPKAPK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK3 is closely related to MAPKAPK2 sharing 72% nucleotide and 75% amino acid identity.
  • Easily Screen and Profile MAPKAPK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human MAPKAPK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK3 is closely related to MAPKAPK2 sharing 72% nucleotide and 75% amino acid identity (1). MAPKAPK3 is activated by growth inducers and stress stimulation of cells. In vitro studies demonstrated that ERK, p38 MAP kinase and Jun N-terminal kinase were all able to phosphorylate and activate this kinase, which suggested the role of MAPKAPK3 as an integrative element of signaling in both mitogen and stress responses (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MAPKAPK3, 10ug (Human, recombinant full-length). MW: ~69kDa. Substrate: HSP27tide (RRLNRQLSVA-amide); derived from the mouse HSP27 (amino acids 80-85). Other: Reaction Buffer, DTT. MAPKAPK3 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/7867/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty