Would you like to log in again?
ADP-Glo Kinase Assay + PKC theta Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4041
PAV4041EA 1140.69 USD InStock
PAV4041
ADP-Glo Kinase Assay + PKC theta Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human PKC theta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein Kinase C theta (PKC theta) is an important component in the intracellular signaling cascade.
  • Easily Screen and Profile PKCθ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PKC theta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein Kinase C theta (PKC theta) is an important component in the intracellular signaling cascade (1). Recent studies have suggested that local accumulation of fat metabolites inside skeletal muscle may activate a serine kinase cascade involving PKC theta leading to defects in insulin signaling and glucose transport in skeletal muscle (2). Insulin resistance plays a primary role in the development of type 2 diabetes and may be related to alterations in fat metabolism. PKC theta is a crucial component mediating fat-induced insulin resistance in skeletal muscle and is a potential therapeutic target for the treatment of type 2 diabetes (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKC theta, 10ug (Human, recombinant full-length). MW: ~110kDa. Substrate: PKCtide (ERMRPRKRQGSVRRRV); derived from protein kinase C epsilon (amino acids 149-164). Other: Reaction Buffer, DTT, Lipid Solution. PKC theta NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5588/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty