Would you like to log in again?
BRK Kinase Enzyme System, Promega
Supplier: Promega
V4054 V6276
PAV4054EA 572.21 USD InStock
PAV4054 PAV6276
BRK Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human BRK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. BRK is a member of the non-receptor tyrosine kinases (PTKs) that contains an amino terminal SH3 and SH2 domain as well as the catalytic domain.
  • Easily Screen and Profile BRK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human BRK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. BRK is a member of the non-receptor tyrosine kinases (PTKs) that contains an amino terminal SH3 and SH2 domain as well as the catalytic domain (1). BRK expression is low or undetectable in normal mammary tissue and benign lesions. However, approximately two-thirds of breast tumors express appreciable levels, and 27% of tumors over express BRK by fivefold or more (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: BRK, 10ug (Human, recombinant full-length). MW: ~80kDa. Substrate: Poly (4:1 Glu, Tyr) peptide. Other: Reaction Buffer, DTT, MnCl2. BRK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5753/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty