Would you like to log in again?
ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4057
PAV4057EA 1140.69 USD InStock
PAV4057
ADP-Glo Kinase Assay + CLK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases.
  • Easily Screen and Profile CLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases. The phosphorylated serine/arginine-rich (SR) proteins are involved in the pre-mRNA processing and releasing through nucleus into nucleoplasm. CLK1, which could phosphorylate the specific SR proteins (1) such as ASF/SF2, may directly regulate the activity and compartmentalization of SR splicing factors (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CLK1, 10ug (Human, recombinant; amino acids 129-end). MW: ~66kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. CLK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1195/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty