Would you like to log in again?
ADP-Glo Kinase Assay + DDR2 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4059
PAV4059EA 1118.32 USD InStock
PAV4059
ADP-Glo Kinase Assay + DDR2 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human DDR2 (467-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. DDR2 is a member of a novel subclass of RTKs containing a distinct extracellular region encompassing a factor VIII-like domain.
  • Easily Screen and Profile DDR2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human DDR2 (467-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. DDR2 is a member of a novel subclass of RTKs containing a distinct extracellular region encompassing a factor VIII-like domain (1) and is thought to be involved in the regulation of cell growth, differentiation and metabolism. DDR2 plays a role in the regulation of collagen turnover mediated by smooth muscle cells in obstructive diseases of blood vessels and the lung (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: DDR2, 10ug (Human, recombinant; amino acids 467-end). MW: ~70kDa. Substrate: Axltide (CKKSRGDYMTMQIG); derived from mouse insulin receptor substrate 1 (amino acids 979-989). Other: Reaction Buffer, DTT, MnCl2. DDR2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4921/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty