Would you like to log in again?
JNK1 Kinase Enzyme System, Promega
Supplier: Promega
V4070 V6440
PAV4070EA 572.31 USD InStock
PAV4070 PAV6440
JNK1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human JNK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JNK1 is a member of the MAP kinase group that is activated by dual phosphorylation at the thr and tyr residues during exposure to stress.
  • Easily Screen and Profile JNK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human JNK1 (sequence matches mouse 100% in protein sequence) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JNK1 is a member of the MAP kinase group that is activated by dual phosphorylation at the thr and tyr residues during exposure to stress such as UV irradiation. JNK1 binds to the c-Jun transactivation domain and phosphorylates it on Ser-63 and Ser-73 (1). JNK1 has been shown to play an important role in disease processes. Activation of JNK1 results in defects in myotube viability and integrity leading to dystrophic myofiber destruction (2). JNK1 activity also is abnormally elevated in obesity, and removal of JNK1 results in decreased adiposity and significantly improved insulin sensitivity. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: JNK1, 10ug (Human, recombinant full-length). MW: ~70kDa. Substrate: p38 substrate (IPTTPITTTYFFFKKK). Other: Reaction Buffer, DTT. JNK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/26419/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty