Would you like to log in again?
ADP-Glo Kinase Assay + MLK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4073
PAV4073EA 1118.32 USD InStock
PAV4073
ADP-Glo Kinase Assay + MLK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human MLK1 (1-433) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Mixed-Lineage Kinase 1 is a mitogen-activated protein kinase kinase kinase capable of activating the c-Jun NH(2)-terminal kinase (JNK) pathway.
  • Easily Screen and Profile MLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human MLK1 (1-433) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK1 or Mixed-Lineage Kinase 1 is a mitogen-activated protein kinase kinase kinase capable of activating the c-Jun NH(2)-terminal kinase (JNK) pathway. The catalytic domain of MLK1 has amino acid sequence similarity to both the tyr-specific and the ser/thr-specific kinase classes. In addition, MLK1 contains 2 leu/ile-zipper motifs and a basic sequence near its C-termini (1). MLK1 is threonine (and possibly serine) phosphorylated at multiple sites in the activation loop, with phosphorylation of threonine 312 required for full activation (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MLK1, 10ug (Human, recombinant; amino acids 1-433). MW: ~77kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. MLK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4293/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty