Would you like to log in again?
ADP-Glo Kinase Assay + p38 delta Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4079
PAV4079EA 1140.69 USD InStock
PAV4079
ADP-Glo Kinase Assay + p38 delta Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human p38 delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. It is a member of the p38 MAPK family and is activated by chemical and environmental stresses as well as by proinflammatory cytokines.
  • Easily Screen and Profile p38δ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human p38 delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p38 delta (SAPK4) is a member of the p38 MAPK family and is activated by chemical and environmental stresses as well as by proinflammatory cytokines. p38 delta has a TGY dual phosphorylation motif and is activated in response to cellular stresses and proinflammatory cytokines (1). MAP kinase kinases 3 and 6 can phosphorylate and activate this kinase. Transcription factor ATF2 and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: p38 delta, 10ug (Human, recombinant full-length). MW: ~71kDa. Substrate: p38 substrate (IPTTPITTTYFFFKKK). Other: Reaction Buffer, DTT. p38 delta NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5603/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty