Would you like to log in again?
STK33 Kinase Enzyme System, Promega
Supplier: Promega
V4086 V6563
PAV6563EA 8648.06 USD InStock
PAV6563 PAV4086
STK33 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human STK33 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. STK33 is a distant member of the CAMK group of serine/threonine kinases.
  • Easily Screen and Profile STK33 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human STK33 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. STK33 is a distant member of the CAMK group of serine/threonine kinases but it lacks the calcium/calmodulin-binding domain and the C-terminal regulatory tail. STK33 is highly expressed in testis, lung epithelia, alveolar macrophages, horizontal cells in the retina and in embryonic organs such as heart, brain and spinal cord. STK33 is essential for abnormal cell growth in human cell lines expressing oncogenic mutations in KRAS but not in human cancer cell lines expressing wildtype KRAS. STK33 is required for survival and proliferation of mutant KRAS-dependent cancer cells in which it suppresses the S6K1-BAD proapoptotic signaling pathway. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: STK33, 10ug (Human, recombinant full-length). MW: ~94kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. STK33 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/65975/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty