Would you like to log in again?
TGF beta R1 Kinase Enzyme System, Promega
Supplier: Promega
V4092 V6566
PAV6566EA 8648.06 USD InStock
PAV6566 PAV4092
TGF beta R1 Kinase Enzyme System, Promega
Assays Enzyme Assays
TGF beta R1 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF beta signal from the cell surface to the cytoplasm.
  • Easily Screen and Profile TGFβR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human TGF beta R1 (ALK5) (80-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TGF beta R1 or transforming growth factor beta-receptor 1 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF beta signal from the cell surface to the cytoplasm. Mutations in the TGF beta R1 gene have been associated with Marfan syndrome, Loeys-Deitz Aortic Aneurysm Syndrome and the development of various types of tumors (1). TGF beta R1-dependent signaling is required for angiogenesis but not for the development of hematopoietic progenitor cells and functional hematopoiesis (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: TGF beta R1, 10ug (Human, recombinant; amino acids 80-end). MW: ~66kDa. Substrate: TGFBR1 Peptide (KKKVLTQMGSPSIRC-S(pS)VS); derived from human SMAD3 (215-230). Other: Reaction Buffer, DTT. TGF beta R1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/7046/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty