Would you like to log in again?
ADP-Glo Kinase Assay + DAPK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4097
PAV4097EA 1140.69 USD InStock
PAV4097
ADP-Glo Kinase Assay + DAPK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human DAPK1 (1-363) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Death-associated protein kinase 1 (DAPK1) is a positive mediator of apoptosis induced by gamma-interferon.
  • Easily Screen and Profile DAPK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human DAPK1 (1-363) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Death-associated protein kinase 1 (DAPK1) is a positive mediator of apoptosis induced by gamma-interferon. Activation of DAPK occurs via dephosphorylation of Ser-308 and subsequent association of calcium/calmodulin (1). DAPK is rapidly dephosphorylated in response to tumor necrosis factor or ceramide and then subsequently degraded via proteasome activity. The decline in DAPK expression is paralleled with increased caspase activity and cell apoptosis. Studies suggest that the apoptosis regulatory activities mediated by DAPK are controlled both by phosphorylation status and protein stability (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: DAPK1, 10ug (Human, recombinant; amino acids 1-363). MW: ~71kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, Ca2+/Calmodulin Solution. DAPK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1612/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty