Would you like to log in again?
ADP-Glo Kinase Assay + SIK Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4157
PAV4157EA 1118.32 USD InStock
PAV4157
ADP-Glo Kinase Assay + SIK Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. SIK is a protein kinase that is involved in regulating AMPK-related kinases.
  • Easily Screen and Profile SIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. SIK is a protein kinase that is involved in regulating AMPK-related kinases (1). SIK may mediate the physiological effects of LKB1, including its tumour suppressor function. SIK is also involved in signaling by various proteins like STRAD, NUAK1, NUAK2, BRSK1, BRSK2, QIK, QSK, SIK, MARK1, MARK2, MARK3, MARK4 and MELK that are related to AMPK. Activation of SIK1 by phosphorylation on Thr322 can lead to an increase in the catalytic activity of sodium/potassium ATPase alpha subunit at the plasma membrane (2). This results in an increase in intracellular sodium in intact mammalian cells. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: SIK, 10ug (Human, recombinant; amino acids 1-303). MW: ~36kDa. Substrate: AMARA peptide (AMARAASAAALARRR). Other: Reaction Buffer, DTT. SIK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/150094/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty