Would you like to log in again?
ADP-Glo Kinase Assay + CLK3 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4163
PAV4163EA 1118.32 USD InStock
PAV4163
ADP-Glo Kinase Assay + CLK3 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human CLK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK3, also known as CDC-like kinase 3, encodes a serine/threonine-type protein kinase with a non-conserved N-terminal domain.
  • Easily Screen and Profile CLK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human CLK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK3, also known as CDC-like kinase 3, encodes a serine/threonine-type protein kinase with a non-conserved N-terminal domain. A long and short isoform (phclk3 and pclk3/152) result from alternative splicing and coexist in different tissues (1). The CLK3 protein has the molecular functions of ATP binding, nucleotide binding, protein serine/threonine kinase activity, protein-tyrosine kinase activity and transferase activity, and the CLK3 protein localizes in both the cytoplasm and nuclear compartment. CLK3 is thought to regulate the intranuclear distribution of the serine/arginine-rich (SR) family of splicing factors. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CLK3, 10ug (Human, recombinant full-length). MW: ~86kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. CLK3 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1198/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty