Would you like to log in again?
MAPKAPK5 Kinase Enzyme System, Promega
Supplier: Promega
V4166 V6445
PAV4166EA 572.31 USD InStock
PAV4166 PAV6445
MAPKAPK5 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human MAPKAPK5 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK5 is a member of the serine/threonine kinase family that responds to cellular stress and proinflammatory cytokines.
  • Easily Screen and Profile MAPKAPK5 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human MAPKAPK5 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK5 is a member of the serine/threonine kinase family that responds to cellular stress and proinflammatory cytokines. MAPKAPK5 is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha and MAPK11/p38-beta (1). MAPKAPK5 is activated in HeLa cells in response to cellular stress and proinflammatory cytokines. MAPKAPK5 activity is regulated by p38-alpha and p38-beta both in vitro and in vivo, and thr-182 is the regulatory phosphorylation site of MAPKAPK5 (2). In vitro, MAPKAPK5 kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: MAPKAPK5, 10ug (Human, recombinant full-length). MW: ~79kDa. Substrate: HSP27tide (RRLNRQLSVA-amide); derived from the mouse HSP27 (amino acids 80-85). Other: Reaction Buffer, DTT. MAPKAPK5 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/8550/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty