Would you like to log in again?
SLK Kinase Enzyme System, Promega
Supplier: Promega
V6562 V4242
PAV4242EA 572.31 USD InStock
PAV4242 PAV6562
SLK Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human SLK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SLK or LOSK is a Ste20-like protein kinase that associates with microtubule and centrosome.
  • Easily Screen and Profile SLK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human SLK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SLK or LOSK is a Ste20-like protein kinase that associates with microtubule and centrosome. Inhibition of SLK activity by dominant-negative mutant or RNAi leads to unfocused microtubule arrangement, indicating that SLK is needed for microtubule organization and proper localization of Golgi complex (1). Expression of v-Src can downregulate SLK activity through the involvement of CK2, which can directly phosphorylate and inhibit SLK activity, whereas inhibition of CK2 in v-Src-transformed cells results in normal SLK activity (2). CK2 and SLK can be co-localized in fibroblasts spreading on fibronectin-coated substrates. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: SLK, 10ug (Human, recombinant full-length). MW: ~180kDa. Substrate: Native Histone H3 Protein; purified from calf thymus tissues. Other: Reaction Buffer, DTT. SLK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/9748/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty