Would you like to log in again?
ADP-Glo Kinase Assay + PRKG1 (PKG) Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4249
PAV4249EA 1196.72 USD InStock
PAV4249
ADP-Glo Kinase Assay + PRKG1 (PKG) Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human PRKG1 (PKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PRKG1 is a homodimer, with each monomer containing a regulatory cGMP-binding domain and a catalytic domain.
  • Easily Screen and Profile PRKG1 (PKG) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human PRKG1 (PKG) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PRKG1 is a homodimer, with each monomer containing a regulatory cGMP-binding domain and a catalytic domain (1). PRKG1 was shown to be expressed at highest levels in bladder, uterus, adrenal gland and fallopian tube. PRKG1 plays an important stimulatory role in platelet activation (2). Expression of recombinant PRKG1 in a reconstituted cell model enhanced von Willebrand factor-induced activation of the platelet integrin alpha-IIb/beta-3. PRKG1 knockout mice showed impaired platelet responses to VWF or low doses of thrombin and prolonged bleeding time. Human platelet aggregation induced by VWF or low-dose thrombin was inhibited by PRKG1 inhibitors but enhanced by cGMP. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PDK1, 10ug (Human, recombinant full-length). MW: ~67kDa. Substrate: PDKtide (KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC); derived from two human proteins: residues 1-14 are based on AKT1 (307-320) and residues 16-39 are based on PKN2/PRK2 (961-984). Other: Reaction Buffer, DTT. PDK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5170/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty