Would you like to log in again?
ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4471
PAV4471EA 1118.32 USD InStock
PAV4471
ADP-Glo Kinase Assay + CAMKK1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
CAMKK1 or CAMKK alpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively.
  • Easily Screen and Profile CAMKK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human CAMKK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMKK1 or CAMKK alpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively. Recent studies have shown that the activity of CAMKK1 is decreased upon phosphorylation by cAMP-dependent protein kinase (PKA) (1). CAMKKI has been identified in intact cells as AMPKKs, predicting a significant role for this kinase in regulating AMPK activity in vivo. It has been shown that 2-deoxyglucose- and ionomycin-stimulated AMPK activity is substantially reduced in HeLa cells transfected with small interfering RNAs specific for CAMKKI (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CAMKK1, 10ug (Human, recombinant full-length). MW: ~94kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, Ca2+/Calmodulin Solution. CAMKK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/84254/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty