Would you like to log in again?
PDGFR Alpha (D842V) Kinase Enzyme System, Promega
Supplier: Promega
V6470 V4480
PAV4480EA 572.31 USD InStock
PAV4480 PAV6470
PDGFR Alpha (D842V) Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human PDGFR alpha (D842V) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
  • Easily Screen and Profile PDGFRα (D842V) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human PDGFR alpha (D842V) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha (platelet-derived growth factor receptor alpha) is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity. Aberrant expression of PDGFR alpha has been linked to developmental abnormalities in vertebrate models and implicated in multiple disease states in humans. There is widespread expression of PDGFR alpha in renal cell types involved in fibrotic and sclerosing processes (1). PDGF and its receptor PDGFR alpha are inducers of fibrosis in the repair phase of inflammatory bowel disease, and they may also be involved in the active inflammatory phase (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PDGFR alpha (D842V), 10ug (Human, recombinant; amino acids 550-end). MW: ~95kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT, MnCl2. PDGFR alpha (D842V) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5156/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty