Would you like to log in again?
ADP-Glo Kinase Assay + CK1 alpha 1 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4485
PAV4485EA 1118.32 USD InStock
PAV4485
ADP-Glo Kinase Assay + CK1 alpha 1 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Full-length recombinant human CK1 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1 alpha 1 is a member of the CK1 family of serine/threonine protein kinases, which play an important role in diverse cell processes.
  • Easily Screen and Profile CK1α1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Full-length recombinant human CK1 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1 alpha 1 is a member of the CK1 family of serine/threonine protein kinases, which play an important role in diverse cell processes. CK1 can regulate Smo cell surface accumulation and activity in response to hedgehog. CK1 phosphorylates Smo at several sites, and phosphorylation-deficient forms of Smo fail to accumulate on the cell surface and are unable to transduce the hedgehog signal (1). CK1 alpha 1 dynamically associates with the CBM complex on T cell receptor engagement to participate in cytokine production and lymphocyte proliferation. CK1 alpha 1 can form complexes with MDM2, which then regulates the stability of p53 and E2F-1 transcription factors (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CK1 alpha 1, 10ug (Human, recombinant full-length). MW: ~62kDa. Substrate: Casein, dephosphorylated; native protein purified from bovine milk. Other: Reaction Buffer, DTT. CK1 alpha 1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1452/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty