Would you like to log in again?
ADP-Glo Kinase Assay + PDGFR alpha (T674I) Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4487
PAV4487EA 1196.72 USD InStock
PAV4487
ADP-Glo Kinase Assay + PDGFR alpha (T674I) Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile PDGFRα (T674I) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human PDGFRα (Y674I) (550-end) point mutant was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFRα (platelet-derived growth factor receptor α) is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity. Aberrant expression of PDGFRα has been linked to developmental abnormalities in vertebrate models and implicated in multiple disease states in humans. There is widespread expression of PDGFRα in renal cell types involved in fibrotic and sclerosing processes. PDGF and its receptor PDGFRα are inducers of fibrosis in the repair phase of inflammatory bowel disease, and they may also be involved in the active inflammatory phase. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PDGFRα (D842V), 10microg (Human, recombinant; amino acids 550-end). MW: ~95kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, MnCl2, DTT. PDGFRα (T674I) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5156/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty