Would you like to log in again?
ALK2 Kinase Enzyme System, Promega
Supplier: Promega
V6272 V4492
PAV6272EA 8648.06 USD InStock
PAV6272 PAV4492
ALK2 Kinase Enzyme System, Promega
Assays Enzyme Assays
ALK2 is a receptor serine/threonine kinase that is member of the ALK family and is upstream of signaling pathway involving the SMAD proteins especially SMAD1/5/8.
  • Easily Screen and Profile ALK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human ALK2 (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ALK2 is a receptor serine/threonine kinase that is member of the ALK family and is upstream of signaling pathway involving the SMAD proteins especially SMAD1/5/8. Knockdown of ALK2, but not TGFbetaRI (ALK5), abrogates endoglin-mediated decrease in cell motility of prostate cancer cells, and constitutively active ALK2 is sufficient to restore a low-motility phenotype in endoglin-deficient cells (1). Therefore, endoglin decreases prostate cancer cell motility through activation of the ALK2-Smad1 pathway. ALK2 is the key gene involved in Fibrodysplasia ossificans progressiva (FOP), a rare autosomal dominant congenital disorder characterized by progressive heterotopic bone formation in muscle tissues (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: ALK2, 10ug (Human, recombinant; amino acids 147-end). MW: ~67kDa. Substrate: Native Casein Protein; purified from bovine milk. Other: Reaction Buffer, DTT. ALK2 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/90/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty