Would you like to log in again?
ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V4501
PAV4501EA 1220.65 USD InStock
PAV4501
ADP-Glo Kinase Assay + NEK3 Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human NEK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a).
  • Easily Screen and Profile NEK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human NEK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a). NEK3 mRNA is detected in all the proliferating cell lines with the amount not changing during the cell cycle. During Prolactin receptor signaling, VAV2 is phosphorylated and activated by NEK3. Overexpression of NEK3 in Chinese hamster ovary cells increases cytoskeletal re-organization in response to Prolactin while downregulation of NEK3 expression by siRNA blocks these effects. Prolactin also stimulates interaction between NEK3 and paxillin leading to increased paxillin phosphorylation. Analysis of breast tissue microarrays show a significant up-regulation of NEK3 expression in malignant versus normal specimens. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: NEK3, 10microg (Human,recombinant full length). MW: ~86kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. NEK3 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/4752/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty