Would you like to log in again?
WNK1 Kinase Enzyme System, Promega
Supplier: Promega
V6599 V5084
PAV6599EA 8648.06 USD InStock
PAV6599 PAV5084
WNK1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human WNK1 (181-507) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile WNK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human WNK1 (181-507) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. WNK1 is a member of the WNK subfamily of serine/threonine protein kinases that is a key regulator of blood pressure by controlling the transport of sodium and chloride ions. Mutations in WNK1 have been associated with pseudohypoaldosteronism type II and hereditary sensory neuropathy type II. WNK1 is a regulator of blood pressure and deficiency of this protein in mice lowers the blood pressure (1). WNK1 can regulate the Ca2+-sensing and the subsequent Ca2+-dependent interactions mediated by synaptotagmin C2 domains, and WNK1 exhibits additive CFTR inhibition. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: WNK1, 10microg (Human, recombinant; amino acids 181-507). MW: ~67kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, MnCl2, DTT. WNK1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/10114/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty