Would you like to log in again?
Trypsin, Gold
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V5280
PAV5280EA 149.81 USD InStock
PAV5280
Trypsin, Gold
Enzymes
Trypsin Gold, Mass Spectrometry Grade, provides maximum specificity for mass spectrometry.
  • Maximum Digest Specificity with Extreme Resistance to Autolytic Digestion
  • Each lot qualified by mass spectrometry to ensures compatibility with your applications/instrumentation
  • Chymotrypsin activity eliminated for distinct, consistent data
  • Digest in gel, or in solution
Trypsin is a serine protease that specifically cleaves at the carboxylic side of lysine and arginine residues. The stringent specificity of trypsin is essential for protein identification. Native trypsin is subject to autolysis, generating pseudotrypsin, which exhibits a broadened specificity including a chymotrypsin-like activity. Such autolysis products, present in a trypsin preparation, would result in additional peptide fragments that could interfere with database analysis of the mass of fragments detected by mass spectrometry. Trypsin Gold, Mass Spectrometry Grade, has been manufactured to provide maximum specificity. Lysine residues in the porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion. The specificity of the purified trypsin is further improved by TPCK treatment, which inactivates chymotrypsin. The treated trypsin is then purified by affinity chromatography and lyophilized to yield Trypsin Gold, Mass Spectrometry Grade. It is resistant to mild denaturing conditions such as 0.1% SDS, 1M urea or 10% acetonitrile and retains 50% of its activity in 2M guanidine HCl. The activity of trypsin is decreased when acidic residues are present on either side of a susceptible bond. If proline is at the carboxylic side of lysine or arginine, the bond is almost completely resistant to cleavage. Each lot of quality-tested Trypsin Gold, Mass Spectrometry Grade, is qualified for use with in-gel digestion and mass spectrometric analysis.
Each lot is quality tested and qualified for use with in-gel digestion and mass spectrometric analysis. The trypsin is provided in one vial, and stability is ensured up to five freeze-thaw cycles.
Native trypsin is subject to autolysis, which broadens its specificity and results in additional peptide fragments that could interfere with database analysis on fragments determined in mass spectrometry. It has been manufactured to provide maximum specificity. Lysine residues in the porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is resistant to autolytic digestion. The specificity is further improved by TPCK treatment to inactivate any chymotrypsin present. The treated trypsin is then purified by affinity chromatography and lyophilized.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty