Would you like to log in again?
AMPK (A1/B1/G2) Kinase Enzyme System, Promega
Supplier: Promega
V4012 V6274
PAV4012EA 1051.14 USD InStock
PAV4012 PAV6274
AMPK (A1/B1/G2) Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as master regulator of cellular energy homeostasis.
  • Easily Screen and Profile AMPK (A1/B1/G2) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeostasis (1). AMPK exists as a heterotrimeric complex composed of a catalytic alpha subunit and regulatory beta and gamma subunits. Binding of AMP to the gamma subunit allosterically activates the complex. AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) (2) and via signaling pathways in response to adiponectin, leptin and CAMKKbeta. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: AMPK (A1/B1/G2), 10ug (Human, recombinant full-length). MW: ~68kDa (A1); ~38kDa (B1); ~65kDa (G2). Substrate: SAMStide (HMRSAMSGLHLVKRR); derived from the mouse acetyl-Coenzyme A carboxylase alpha (amino acids 73-85). Other: Reaction Buffer, DTT, AMP Solution. AMPK (A1/B1/G2) NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5562/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty