Would you like to log in again?
CAMK1 gamma Kinase Enzyme System, Promega
Supplier: Promega
V6277 V4016
PAV6277EA 8648.06 USD InStock
PAV6277 PAV4016
CAMK1 gamma Kinase Enzyme System, Promega
Assays Enzyme Assays
Full-length recombinant human CAMK1 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK1 gamma (CLICK-III), a member of the CAMK family, is a novel membrane-anchored neuronal Ca2+/calmodulin-dependent protein kinase.
  • Easily Screen and Profile CAMK1γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Full-length recombinant human CAMK1 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK1 gamma (CLICK-III), a member of the CAMK family, is a novel membrane-anchored neuronal Ca2+/calmodulin-dependent protein kinase. Full activation of CaMK1 gamma requires both Ca2+/CaM and phosphorylation by CAMKK. CAMK1 gamma transcript is most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus (1). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CAMK1 gamma, 10ug (Human, recombinant full-length). MW: ~80kDa. Substrate: Autocamtide 2 peptide (KKALRRQETVDAL-amide); derived from the autophosphorylation site (amino acids 283-290) on CaMKII. Other: Reaction Buffer, DTT, Ca2+/Calmodulin Solution. CAMK1 gamma NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/57172/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty