Would you like to log in again?
FMS Kinase Enzyme System, Promega
Supplier: Promega
V4022 V6395
PAV4022EA 583.65 USD InStock
PAV4022 PAV6395
FMS Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R).
  • Easily Screen and Profile FMS Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FMS is a proto-oncogene that encodes the tyrosine kinase transmembrane receptor for colony stimulating factor 1 receptor (CSF1R). FMS is homodimeric, contains a so-called kinase insert domain and is a member of the CSF1R/PDGF receptor family of tyrosine-protein kinases. FMS mediates most if not all of the biological effects of CSF1, which control the production, differentiation and cell function the monocyte/macrophage lineage (1). Mutations in FMS have been associated with providing sustained signals for cell growth and a predisposition to myeloid malignancy (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: FMS, 10ug (Human, recombinant; amino acids 539-end). MW: ~76kDa. Substrate: Poly (4:1 Glu, Tyr) peptide. Other: Reaction Buffer, DTT, MnCl2. FMS NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1436/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty