Would you like to log in again?
IKK alpha Kinase Enzyme System, Promega
Supplier: Promega
V4068 V6399
PAV6399EA 8648.06 USD InStock
PAV6399 PAV4068
IKK alpha Kinase Enzyme System, Promega
Assays Enzyme Assays
Full-length recombinant human IKK alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. IKK alpha is a serine/threonine protein kinase that phosphorylates the I-kappa-B protein.
  • Easily Screen and Profile IKKα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Full-length recombinant human IKK alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. IKK alpha is a serine/threonine protein kinase that phosphorylates the I-kappa-B protein, which is an inhibitor of the transcription factor NF-kappa-B complex. Phosphorylation of I-kappa-B protein triggers the degradation of the inhibitor via the ubiquitination pathway, thereby activating NF-kappa-B complex (1). IKK alpha is an essential regulator of NF-kappa-B-dependent gene expression through control of promoter-associated histone phosphorylation after cytokine exposure (2). IKK alpha is a critical component of the cytoplasmic transductional-transcriptional processor leading to induction of IFN alpha production. IKK alpha is also involved in the epidermis, where it antagonizes mitogenic and angiogenic signals and represses tumor progression and metastases. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: IKK alpha, 10ug (Human, recombinant full-length). MW: ~114kDa. Substrate: IKKtide (KKKKERLLDDRHDSG-LDSMKDEE); derived from human IkBA (amino acids 21-41). Other: Reaction Buffer, DTT. IKK alpha NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/1147/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty